Product Details

SNP ID
rs200419059
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:201040255 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTTTGGATCAGCAGGGCTGTAGC[A/T]GGTGCTGAGCACCTGGAAGTATTCT
Phenotype
MIM: 114208
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CACNA1S PubMed Links

Gene Details

Gene
CACNA1S
Gene Name
calcium voltage-gated channel subunit alpha1 S
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000069.2 5573 Silent Mutation CCA,CCT P1782P NP_000060.2
XM_005245478.3 5573 Silent Mutation CCA,CCT P1763P XP_005245535.1
Gene
LOC101929305
Gene Name
uncharacterized LOC101929305
There are no transcripts associated with this gene.

View Full Product Details