Product Details

SNP ID
rs202036389
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:83870031 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCATTCCAGGTGAATTGTACTTCAA[A/G]TTGTAGGTTGGTGTTCTCAAGAAGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC101927560 PubMed Links

Gene Details

Gene
LOC101927560
Gene Name
uncharacterized LOC101927560
There are no transcripts associated with this gene.

Gene
LOC101927587
Gene Name
uncharacterized LOC101927587
There are no transcripts associated with this gene.

Gene
TTLL7
Gene Name
tubulin tyrosine ligase like 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024686.4 2766 Silent Mutation AAC,AAT N865N NP_078962.4
XM_005271208.2 2766 Silent Mutation AAC,AAT N838N XP_005271265.1
XM_011542159.2 2766 Intron XP_011540461.1
XM_017002347.1 2766 Silent Mutation AAC,AAT N865N XP_016857836.1
XM_017002348.1 2766 Intron XP_016857837.1
XM_017002349.1 2766 Silent Mutation AAC,AAT N620N XP_016857838.1

View Full Product Details