Product Details

SNP ID
rs202235631
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:23750937 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGAAGGAGGAGGAGATGGAGGGGG[A/G]CTACCAAGAAACCTGGAAAGCCACG
Phenotype
MIM: 600786
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TCEB3 PubMed Links

Gene Details

Gene
TCEB3
Gene Name
transcription elongation factor B subunit 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003198.2 470 Missense Mutation GAC,GGC D137G NP_003189.2
Gene
TCEB3-AS1
Gene Name
TCEB3 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details