Product Details

SNP ID
rs202145940
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:206866612 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGCCTCGCCAATTCCTTTCTTAC[C/G]ATCAAGAAGGACCTCCGGCTCTGTG
Phenotype
MIM: 605619
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
IL20 PubMed Links

Gene Details

Gene
IL20
Gene Name
interleukin 20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018724.3 824 Silent Mutation ACC,ACG T118T NP_061194.2
XM_005273152.3 824 Silent Mutation ACC,ACG T203T XP_005273209.1

View Full Product Details