Product Details

SNP ID
rs199540085
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:36879069 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTTTCACTTCCCACCAAGAGTCA[C/T]CGGCCATCCCTGGAGTCTGGTCTTC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TLDC2 PubMed Links

Gene Details

Gene
TLDC2
Gene Name
TBC/LysM-associated domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304783.1 112 Missense Mutation ACC,ATC T73I NP_001291712.1
NM_080628.2 112 Missense Mutation ACC,ATC T73I NP_542195.1
XM_017027672.1 112 Missense Mutation ACC,ATC T73I XP_016883161.1
XM_017027673.1 112 Missense Mutation ACC,ATC T73I XP_016883162.1
XM_017027674.1 112 UTR 5 XP_016883163.1

View Full Product Details