Product Details

SNP ID
rs200815842
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:44504806 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTAAATAATACTGTTCACCGAGAA[A/G]GTACGCCCATCGAAGGAGTTGGCCA
Phenotype
MIM: 612920
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TSPEAR PubMed Links

Gene Details

Gene
TSPEAR
Gene Name
thrombospondin type laminin G domain and EAR repeats
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001272037.1 1958 Silent Mutation ACC,ACT T542T NP_001258966.1
NM_144991.2 1958 Silent Mutation ACC,ACT T610T NP_659428.2
Gene
TSPEAR-AS1
Gene Name
TSPEAR antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details