Product Details
- SNP ID
-
rs200670934
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:16592229 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGCAAGGCTTCCGTCAGCAGAACCA[C/T]GAAGGCTGTGCCGTCCCCACTATTC
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CCT8L2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs5747988] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CCT8L2
- Gene Name
- chaperonin containing TCP1 subunit 8 like 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_014406.4 |
|
Intron |
|
|
NP_055221.1 |
- Gene
- TPTEP1
- Gene Name
- transmembrane phosphatase with tensin homology pseudogene 1
There are no transcripts associated with this gene.
View Full Product Details