Product Details

SNP ID
rs201986123
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:37824862 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCTACTACGGCACCGTGCGCTAC[A/G]GCGCGCTGGAGCTCTGCGTGCCCGC
Phenotype
MIM: 613383 MIM: 603692 MIM: 607422
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANKRD54 PubMed Links

Gene Details

Gene
ANKRD54
Gene Name
ankyrin repeat domain 54
There are no transcripts associated with this gene.

Gene
GALR3
Gene Name
galanin receptor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003614.1 524 Missense Mutation AGC,GGC S167G NP_003605.1
Gene
GCAT
Gene Name
glycine C-acetyltransferase
There are no transcripts associated with this gene.

View Full Product Details