Product Details

SNP ID
rs201693188
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:50482623 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCAGCCCAGGCACCCCGCACCCC[A/G]ACCTGTGGTCTGGAAGCTTCACCGG
Phenotype
MIM: 608682 MIM: 606774 MIM: 603560
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADM2 PubMed Links

Gene Details

Gene
ADM2
Gene Name
adrenomedullin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001253845.1 218 Missense Mutation CAA,CGA Q56R NP_001240774.1
XM_017028940.1 218 Missense Mutation CAA,CGA Q56R XP_016884429.1
Gene
MIOX
Gene Name
myo-inositol oxygenase
There are no transcripts associated with this gene.

Gene
SBF1
Gene Name
SET binding factor 1
There are no transcripts associated with this gene.

View Full Product Details