Product Details

SNP ID
rs199992222
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:25351898 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCATTCCTGTCCGCATGAGGGTT[C/G]GTTCCTGGAAGTGAACACACAGGCT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LOC100652901 PubMed Links

Gene Details

Gene
LOC100652901
Gene Name
uncharacterized LOC100652901
There are no transcripts associated with this gene.

Gene
LRP5L
Gene Name
LDL receptor related protein 5 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135772.1 3786 Silent Mutation ACC,ACG T220T NP_001129244.1
NM_182492.2 3786 Intron NP_872298.1
XM_006724372.3 3786 Intron XP_006724435.1
XM_017029083.1 3786 Silent Mutation ACC,ACG T220T XP_016884572.1
XM_017029084.1 3786 Silent Mutation ACC,ACG T220T XP_016884573.1
XM_017029085.1 3786 Silent Mutation ACC,ACG T220T XP_016884574.1
XM_017029086.1 3786 Silent Mutation ACC,ACG T220T XP_016884575.1
XM_017029087.1 3786 Silent Mutation ACC,ACG T220T XP_016884576.1
XM_017029088.1 3786 Intron XP_016884577.1

View Full Product Details