Product Details

SNP ID
rs200343776
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:50481895 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTGGGTTGCATCAGCCTCCTCTG[C/G]CTGCAGCTCCCTGGCTCGCTGTCCC
Phenotype
MIM: 608682 MIM: 606774 MIM: 603560
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ADM2 PubMed Links

Gene Details

Gene
ADM2
Gene Name
adrenomedullin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001253845.1 99 Missense Mutation TGC,TGG C16W NP_001240774.1
XM_017028940.1 99 Missense Mutation TGC,TGG C16W XP_016884429.1
Gene
MIOX
Gene Name
myo-inositol oxygenase
There are no transcripts associated with this gene.

Gene
SBF1
Gene Name
SET binding factor 1
There are no transcripts associated with this gene.

View Full Product Details