Product Details

SNP ID
rs201237992
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:196646381 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGTGTCCAGACCGGTCCTTTCTC[C/T]AACCCACATCAACGCTACAGCTTCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC150 PubMed Links

Gene Details

Gene
CCDC150
Gene Name
coiled-coil domain containing 150
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080539.1 188 Missense Mutation CCA,CTA P18L NP_001074008.1
XM_006712438.3 188 Missense Mutation CCA,CTA P18L XP_006712501.1
XM_006712439.3 188 Intron XP_006712502.1
XM_011510985.2 188 Intron XP_011509287.1
XM_011510987.2 188 Intron XP_011509289.1
XM_011510988.2 188 Intron XP_011509290.1
XM_017003861.1 188 Missense Mutation CCA,CTA P18L XP_016859350.1
XM_017003862.1 188 Missense Mutation CCA,CTA P18L XP_016859351.1
XM_017003863.1 188 Missense Mutation CCA,CTA P18L XP_016859352.1
XM_017003864.1 188 Missense Mutation CCA,CTA P18L XP_016859353.1
XM_017003865.1 188 Intron XP_016859354.1
XM_017003866.1 188 Intron XP_016859355.1

View Full Product Details