Product Details

SNP ID
rs200971045
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:200856731 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATGTCTTGTAGATACCAATGTACT[A/G]TGATGTTCGTCATCATATGTTGCAC
Phenotype
MIM: 601951
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CLK1 PubMed Links

Gene Details

Gene
CLK1
Gene Name
CDC like kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001162407.1 1189 Silent Mutation CAC,CAT H378H NP_001155879.1
NM_004071.3 1189 Silent Mutation CAC,CAT H336H NP_004062.2

View Full Product Details