Product Details

SNP ID
rs199667955
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:161995029 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCTGAGCTGACTGCTGAAAGTGAA[C/T]GTTATCTGCAGGGAGAGAAAGGAAA
Phenotype
MIM: 102720 MIM: 605556
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DPP4 PubMed Links

Gene Details

Gene
DPP4
Gene Name
dipeptidyl peptidase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001935.3 2692 Missense Mutation ATT,GTT I711V NP_001926.2
XM_005246371.2 2692 Missense Mutation ATT,GTT I710V XP_005246428.1
Gene
SLC4A10
Gene Name
solute carrier family 4 member 10
There are no transcripts associated with this gene.

View Full Product Details