Product Details

SNP ID
rs200529878
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:47373902 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGGGCCCTCCAGAACAATGATGG[A/G]CTTTATGATCCTGACTGCGATGAGA
Phenotype
MIM: 185535
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EPCAM PubMed Links

Gene Details

Gene
EPCAM
Gene Name
epithelial cell adhesion molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002354.2 637 Silent Mutation GGA,GGG G93G NP_002345.2
Gene
MIR559
Gene Name
microRNA 559
There are no transcripts associated with this gene.

View Full Product Details