Product Details

SNP ID
rs200488482
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:240869003 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCAGCCGGGGGGCTGCAGATGAT[C/T]GGGTCCATGAGCAAGGATATGTACC
Phenotype
MIM: 604285
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AGXT PubMed Links

Gene Details

Gene
AGXT
Gene Name
alanine-glyoxylate aminotransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000030.2 259 Silent Mutation ATC,ATT I46I NP_000021.1

View Full Product Details