Product Details

SNP ID
rs199728784
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:61483060 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGCTGCAGCTTCTTCTTGTGCAAC[A/G]TTTTGTAAGAGTGTAAAAAGTATCT
Phenotype
MIM: 602559
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
XPO1 PubMed Links

Gene Details

Gene
XPO1
Gene Name
exportin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003400.3 3437 Silent Mutation AAC,AAT N903N NP_003391.1
XM_005264544.1 3437 Silent Mutation AAC,AAT N888N XP_005264601.1
XM_005264546.1 3437 Silent Mutation AAC,AAT N772N XP_005264603.1
XM_006712094.2 3437 Silent Mutation AAC,AAT N903N XP_006712157.1
XM_011533097.1 3437 Silent Mutation AAC,AAT N903N XP_011531399.1
XM_011533098.1 3437 Silent Mutation AAC,AAT N858N XP_011531400.1
XM_011533099.2 3437 Silent Mutation AAC,AAT N837N XP_011531401.1

View Full Product Details