Product Details

SNP ID
rs202128623
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:199956183 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGGTTAGGACTCGAATTAAGGCC[C/T]TCCTTATCTGGGCCTATTTCGACAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C2orf47 PubMed Links

Gene Details

Gene
C2orf47
Gene Name
chromosome 2 open reading frame 47
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024520.2 618 Missense Mutation CTC,TTC L129F NP_078796.2
XM_005246852.2 618 Missense Mutation CTC,TTC L129F XP_005246909.1
Gene
TYW5
Gene Name
tRNA-yW synthesizing protein 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039693.2 618 Intron NP_001034782.1
XM_017003310.1 618 Intron XP_016858799.1

View Full Product Details