Product Details

SNP ID
rs202066629
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:26191525 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGCCAAGCCCAAAGACGGCTCCGA[C/T]GTCTCCCTCTGCAGGTGTGGCCAAG
Phenotype
MIM: 600890
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GAREM2 PubMed Links

Gene Details

Gene
GAREM2
Gene Name
GRB2 associated regulator of MAPK1 subtype 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001168241.1 2234 Intron NP_001161713.1
NM_001191033.1 2234 Intron NP_001177962.1
XM_006711951.3 2234 Intron XP_006712014.1
XM_011532564.2 2234 Intron XP_011530866.1
XM_011532565.2 2234 Intron XP_011530867.1
XM_011532566.2 2234 Intron XP_011530868.1
XM_011532567.2 2234 Intron XP_011530869.1
Gene
HADHA
Gene Name
hydroxyacyl-CoA dehydrogenase/3-ketoacyl-CoA thiolase/enoyl-CoA hydratase (trifunctional protein), alpha subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000182.4 2234 Missense Mutation NP_000173.2

View Full Product Details