Product Details

SNP ID
rs202190979
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:151213416 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTTACATAAGATTTCCCTAAACG[A/G]CTGGCATAGAAAGAAATAAATAATA
Phenotype
MIM: 611318 MIM: 610116
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GPR171 PubMed Links

Gene Details

Gene
GPR171
Gene Name
G protein-coupled receptor 171
There are no transcripts associated with this gene.

Gene
MED12L
Gene Name
mediator complex subunit 12 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_053002.5 1189 Intron NP_443728.3
XM_006713487.3 1189 Intron XP_006713550.1
XM_011512390.2 1189 Intron XP_011510692.1
XM_011512394.2 1189 Intron XP_011510696.1
XM_011512399.2 1189 Intron XP_011510701.1
XM_017005676.1 1189 Intron XP_016861165.1
XM_017005677.1 1189 Intron XP_016861166.1
XM_017005678.1 1189 Intron XP_016861167.1
XM_017005679.1 1189 Intron XP_016861168.1
XM_017005680.1 1189 Intron XP_016861169.1
XM_017005681.1 1189 Intron XP_016861170.1
Gene
P2RY14
Gene Name
purinergic receptor P2Y14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001081455.1 1189 Missense Mutation NP_001074924.1
NM_014879.3 1189 Missense Mutation NP_055694.3
XM_005247922.3 1189 Missense Mutation XP_005247979.1
XM_005247923.3 1189 Missense Mutation XP_005247980.1
XM_011513340.2 1189 Missense Mutation XP_011511642.1
XM_017007583.1 1189 Missense Mutation XP_016863072.1

View Full Product Details