Product Details

SNP ID
rs201922651
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:52231154 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTTAATGCGGATCTGCTGGAGCTC[C/T]CTCTCAGCCGAGGTCAGCGGGGCAG
Phenotype
MIM: 605474 MIM: 607433
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC101929054 PubMed Links

Gene Details

Gene
LOC101929054
Gene Name
uncharacterized LOC101929054
There are no transcripts associated with this gene.

Gene
TLR9
Gene Name
toll like receptor 9
There are no transcripts associated with this gene.

Gene
TWF2
Gene Name
twinfilin actin binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007284.3 607 Silent Mutation AGA,AGG R152R NP_009215.1

View Full Product Details