Product Details

SNP ID
rs201677530
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.3:27717254 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTGAATCATTGGAATCTAGAGAT[G/T]TGATGGAAGGGGGTGTCTCTATCCA
Phenotype
MIM: 604615
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
EOMES PubMed Links

Gene Details

Gene
EOMES
Gene Name
eomesodermin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278182.1 2234 Missense Mutation AAA,ACA K645T NP_001265111.1
NM_001278183.1 2234 Missense Mutation AAA,ACA K350T NP_001265112.1
NM_005442.3 2234 Missense Mutation AAA,ACA K626T NP_005433.2
XM_005265510.3 2234 UTR 3 XP_005265567.1

View Full Product Details