Product Details

SNP ID
rs201179424
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:39265634 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAACTTCTCCCCAGCAAATGCATA[C/G]ATGAGAGGATTCAGGCAACAATGGC
Phenotype
MIM: 601470
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CX3CR1 PubMed Links

Gene Details

Gene
CX3CR1
Gene Name
C-X3-C motif chemokine receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171171.1 968 Missense Mutation ATC,ATG I292M NP_001164642.1
NM_001171172.1 968 Missense Mutation ATC,ATG I292M NP_001164643.1
NM_001171174.1 968 Missense Mutation ATC,ATG I324M NP_001164645.1
NM_001337.3 968 Missense Mutation ATC,ATG I292M NP_001328.1

View Full Product Details