Product Details

SNP ID
rs200203667
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:125530572 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCAAAGTGGTTTATGATAAACCCA[G/T]CCATCTCCCTGAAACAAATAAAAAG
Phenotype
MIM: 606739
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
OSBPL11 PubMed Links

Gene Details

Gene
OSBPL11
Gene Name
oxysterol binding protein like 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022776.4 2924 Silent Mutation GGA,GGC G729G NP_073613.2

View Full Product Details