Product Details

SNP ID
rs199840845
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:69589481 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCACACAGACCAGCAAGAACCCA[A/C]TTACATCCAAAGAGTGGTACTGGAA
Phenotype
MIM: 604716
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
UGT2A1 PubMed Links

Gene Details

Gene
UGT2A1
Gene Name
UDP glucuronosyltransferase family 2 member A1 complex locus
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001252274.2 1372 Missense Mutation AGT,ATT S658I NP_001239203.2
NM_001252275.2 1372 Missense Mutation AGT,ATT S492I NP_001239204.2
NM_001301239.1 1372 Missense Mutation AGT,ATT S448I NP_001288168.1
NM_006798.4 1372 Missense Mutation AGT,ATT S492I NP_006789.3
Gene
UGT2A2
Gene Name
UDP glucuronosyltransferase family 2 member A2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001105677.2 1372 Missense Mutation AGT,ATT S501I NP_001099147.2
NM_001301233.1 1372 Missense Mutation AGT,ATT S457I NP_001288162.1

View Full Product Details