Product Details

SNP ID
rs199568047
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:183510707 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCAAGGCCAAGCAGGAAAGGGAA[G/T]CTTCACCTGTTGAGTTTGCAATAGA
Phenotype
MIM: 604215
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ING2 PubMed Links

Gene Details

Gene
ING2
Gene Name
inhibitor of growth family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291959.1 589 Missense Mutation GCT,TCT A160S NP_001278888.1
NM_001564.3 589 Missense Mutation GCT,TCT A200S NP_001555.1
XM_011531927.2 589 Missense Mutation GCT,TCT A145S XP_011530229.1
Gene
LOC389247
Gene Name
uncharacterized LOC389247
There are no transcripts associated with this gene.

View Full Product Details