Product Details
- SNP ID
-
rs199579283
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:154863386 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAGAAATCGTGCTCAGTTACAGTTA[C/T]ATTGCCATGGTAAGGAGCTCTACTC
- Phenotype
-
MIM: 603731
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CNOT8
PubMed Links
Gene Details
- Gene
- CNOT8
- Gene Name
- CCR4-NOT transcription complex subunit 8
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001301073.1 |
430 |
Silent Mutation |
TAC,TAT |
Y36Y |
NP_001288002.1 |
NM_001301074.1 |
430 |
Silent Mutation |
TAC,TAT |
Y36Y |
NP_001288003.1 |
NM_001301075.1 |
430 |
UTR 5 |
|
|
NP_001288004.1 |
NM_001301077.1 |
430 |
Intron |
|
|
NP_001288006.1 |
NM_001301080.1 |
430 |
Intron |
|
|
NP_001288009.1 |
NM_001301082.1 |
430 |
Intron |
|
|
NP_001288011.1 |
NM_001301083.1 |
430 |
Intron |
|
|
NP_001288012.1 |
NM_001301086.1 |
430 |
Intron |
|
|
NP_001288015.1 |
NM_004779.5 |
430 |
Silent Mutation |
TAC,TAT |
Y36Y |
NP_004770.4 |
XM_005268527.2 |
430 |
Silent Mutation |
TAC,TAT |
Y36Y |
XP_005268584.1 |
XM_011537706.1 |
430 |
UTR 5 |
|
|
XP_011536008.1 |
XM_011537708.2 |
430 |
Intron |
|
|
XP_011536010.1 |
XM_017010048.1 |
430 |
Silent Mutation |
TAC,TAT |
Y36Y |
XP_016865537.1 |
XM_017010049.1 |
430 |
Silent Mutation |
TAC,TAT |
Y36Y |
XP_016865538.1 |
XM_017010050.1 |
430 |
Silent Mutation |
TAC,TAT |
Y36Y |
XP_016865539.1 |
XM_017010051.1 |
430 |
Silent Mutation |
TAC,TAT |
Y36Y |
XP_016865540.1 |
XM_017010052.1 |
430 |
Silent Mutation |
TAC,TAT |
Y36Y |
XP_016865541.1 |
XM_017010053.1 |
430 |
Silent Mutation |
TAC,TAT |
Y36Y |
XP_016865542.1 |
XM_017010054.1 |
430 |
Silent Mutation |
TAC,TAT |
Y36Y |
XP_016865543.1 |
XM_017010055.1 |
430 |
Silent Mutation |
TAC,TAT |
Y36Y |
XP_016865544.1 |
XM_017010056.1 |
430 |
Silent Mutation |
TAC,TAT |
Y36Y |
XP_016865545.1 |
XM_017010057.1 |
430 |
UTR 5 |
|
|
XP_016865546.1 |
XM_017010058.1 |
430 |
Intron |
|
|
XP_016865547.1 |
View Full Product Details