Product Details

SNP ID
rs200621359
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:147881457 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTTTTCCTGCAGCTACTGCCTTC[C/G]TCATCAACAAAGTGCCCCTTCCTGT
Phenotype
MIM: 606531
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
C5orf46 PubMed Links

Gene Details

Gene
C5orf46
Gene Name
chromosome 5 open reading frame 46
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_206966.2 160 Intron NP_996849.2
XM_005268446.3 160 Intron XP_005268503.2
XM_017009459.1 160 Intron XP_016864948.1
Gene
SCGB3A2
Gene Name
secretoglobin family 3A member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_054023.4 160 Missense Mutation CTC,GTC L23V NP_473364.1

View Full Product Details