Product Details

SNP ID
rs200769603
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:157138956 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAATTCTGAATCATTTCAATTACT[C/T]GTTCCAGGTGCTTTTGAAATCTCTC
Phenotype
MIM: 605045
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MED7 PubMed Links

Gene Details

Gene
MED7
Gene Name
mediator complex subunit 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001100816.1 584 Missense Mutation CAA,CGA Q159R NP_001094286.1
NM_004270.4 584 Missense Mutation CAA,CGA Q159R NP_004261.1

View Full Product Details