Product Details

SNP ID
rs200915022
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:90518963 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGATGGGGGTGTGATTTTTGAATG[C/T]AAATGTGAAGAGTCCACTGTTGAAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LYSMD3 PubMed Links

Gene Details

Gene
LYSMD3
Gene Name
LysM domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286812.1 1390 UTR 3 NP_001273741.1
NM_198273.2 1390 Silent Mutation TTA,TTG L259L NP_938014.1
XM_005248421.4 1390 Silent Mutation TTA,TTG L259L XP_005248478.1
Gene
POLR3G
Gene Name
RNA polymerase III subunit G
There are no transcripts associated with this gene.

View Full Product Details