Product Details

SNP ID
rs200638839
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:132658294 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGGCGGCTTTGCCTCCCCAGGCCC[C/T]GTGCCTCCCTCTACAGCCCTCAGGG
Phenotype
MIM: 147683
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IL13 PubMed Links

Gene Details

Gene
IL13
Gene Name
interleukin 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002188.2 122 Silent Mutation CCC,CCT P36P NP_002179.2
Gene
TH2LCRR
Gene Name
T helper type 2 locus control region associated RNA
There are no transcripts associated with this gene.

View Full Product Details