Product Details

SNP ID
rs202081635
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:137493768 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTCCAGGGAGCTGGTGAGCATGA[A/G]GATGTAGTTTCTGGCGAGCAGGAGT
Phenotype
MIM: 609323
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OLIG3 PubMed Links

Gene Details

Gene
OLIG3
Gene Name
oligodendrocyte transcription factor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175747.2 627 Missense Mutation CTC,TTC L135F NP_786923.1

View Full Product Details