Product Details

SNP ID
rs202088854
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:46799116 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTTGTTTTCACAGAAATCCAGAA[A/G]TGGCCTGAGAGACCCAAACACCAGG
Phenotype
MIM: 600388
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MEP1A PubMed Links

Gene Details

Gene
MEP1A
Gene Name
meprin A subunit alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005588.2 239 Missense Mutation AAT,AGT N66S NP_005579.2
XM_011514628.1 239 Missense Mutation AAT,AGT N94S XP_011512930.1
XM_011514629.1 239 Missense Mutation AAT,AGT N66S XP_011512931.1

View Full Product Details