Product Details

SNP ID
rs200865884
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:25726391 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCGCCAAATACACTGGTGCGCCT[A/G]CCCCTATCCGCTCTGCATAGTTTCC
Phenotype
MIM: 613499 MIM: 609904 MIM: 182308
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
HIST1H2AA PubMed Links
Additional Information
For this assay, SNP(s) [rs9467583] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
HIST1H2AA
Gene Name
histone cluster 1, H2aa
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_170745.3 172 Missense Mutation GCA,GTA A46V NP_734466.1
Gene
HIST1H2APS1
Gene Name
histone cluster 1, H2a, pseudogene 1
There are no transcripts associated with this gene.

Gene
HIST1H2BA
Gene Name
histone cluster 1, H2ba
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_170610.2 172 Intron NP_733759.1
Gene
SLC17A1
Gene Name
solute carrier family 17 member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005074.3 172 Intron NP_005065.2
XM_011514818.2 172 Intron XP_011513120.2
XM_011514819.2 172 Intron XP_011513121.2
XM_011514820.2 172 Intron XP_011513122.2
XM_011514821.2 172 Intron XP_011513123.1
XM_017011199.1 172 Intron XP_016866688.1
XM_017011200.1 172 Intron XP_016866689.1
XM_017011201.1 172 Intron XP_016866690.1
XM_017011202.1 172 Intron XP_016866691.1

View Full Product Details