Product Details

SNP ID
rs201466479
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:129833500 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAACATGAAGAATAGTGACTGAAAT[A/G]GCATAATCCCAAACCCATTCTTCCA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TMEM244 PubMed Links

Gene Details

Gene
TMEM244
Gene Name
transmembrane protein 244
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010876.1 535 Silent Mutation GCC,GCT A93A NP_001010876.1
XM_011535669.2 535 Silent Mutation GCC,GCT A93A XP_011533971.1

View Full Product Details