Product Details

SNP ID
rs199559364
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:27132672 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGCGGCTGCGCTTGCGCTTCTTGC[C/T]GTCTTTCTTCTGCGCCTTAGTCACC
Phenotype
MIM: 615012 MIM: 615044 MIM: 615045 MIM: 602833
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HIST1H2AG PubMed Links

Gene Details

Gene
HIST1H2AG
Gene Name
histone cluster 1, H2ag
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021064.4 125 Intron NP_066408.1
Gene
HIST1H2BJ
Gene Name
histone cluster 1, H2bj
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021058.3 125 Missense Mutation AGC,GGC S27G NP_066402.2
Gene
HIST1H2BK
Gene Name
histone cluster 1, H2bk
There are no transcripts associated with this gene.

Gene
HIST1H4I
Gene Name
histone cluster 1, H4i
There are no transcripts associated with this gene.

View Full Product Details