Product Details

SNP ID
rs200645947
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:23506143 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTCAAGTTAAAACATTACCTGTA[A/T]CGGTAATCATAGTCTTCATATCTGT
Phenotype
MIM: 602718
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
TRA2A PubMed Links

Gene Details

Gene
TRA2A
Gene Name
transformer 2 alpha homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282757.1 Intron NP_001269686.1
NM_001282758.1 Intron NP_001269687.1
NM_001282759.1 Intron NP_001269688.1
NM_013293.4 Intron NP_037425.1
XM_005249725.1 Intron XP_005249782.1
XM_011515331.1 Intron XP_011513633.1
XM_017012089.1 Intron XP_016867578.1
XM_017012090.1 Intron XP_016867579.1

View Full Product Details