Product Details

SNP ID
rs200783579
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:21902983 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGCTAGAGACTTACTTACCTCTC[C/T]AGATATTCCTTAACATTGTCATAAC
Phenotype
MIM: 609685 MIM: 603339
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CDCA7L PubMed Links

Gene Details

Gene
CDCA7L
Gene Name
cell division cycle associated 7 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127370.2 1449 Silent Mutation CTA,CTG L409L NP_001120842.1
NM_001127371.2 1449 Silent Mutation CTA,CTG L397L NP_001120843.1
NM_018719.4 1449 Silent Mutation CTA,CTG L443L NP_061189.2
Gene
DNAH11
Gene Name
dynein axonemal heavy chain 11
There are no transcripts associated with this gene.

View Full Product Details