Product Details

SNP ID
rs200466467
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:21901012 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGCTGTGTTTTTGCCATAGGCGCC[C/T]GCTGGGACACCCAAGCAGGAACCAT
Phenotype
MIM: 609685 MIM: 603339
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CDCA7L PubMed Links

Gene Details

Gene
CDCA7L
Gene Name
cell division cycle associated 7 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127370.2 2795 UTR 3 NP_001120842.1
NM_001127371.2 2795 UTR 3 NP_001120843.1
NM_018719.4 2795 UTR 3 NP_061189.2
Gene
DNAH11
Gene Name
dynein axonemal heavy chain 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001277115.1 2795 Missense Mutation CGC,TGC R4437C NP_001264044.1

View Full Product Details