Product Details

SNP ID
rs201046984
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:140523397 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTTCCAGGGCCACCTGAAGCTTCC[A/G]GGGCAGGATGGAGTCCTCATCGTCC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DENND2A PubMed Links

Gene Details

Gene
DENND2A
Gene Name
DENN domain containing 2A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318052.1 3077 Missense Mutation CGG,TGG R859W NP_001304981.1
NM_001318053.1 3077 Intron NP_001304982.1
NM_015689.4 3077 Missense Mutation CGG,TGG R859W NP_056504.3
XM_005249977.3 3077 Missense Mutation CGG,TGG R859W XP_005250034.1
XM_005249979.4 3077 Intron XP_005250036.1
XM_011516052.2 3077 Missense Mutation CGG,TGG R859W XP_011514354.1
XM_011516053.2 3077 Missense Mutation CGG,TGG R859W XP_011514355.1
XM_011516054.2 3077 Missense Mutation CGG,TGG R858W XP_011514356.1
XM_017011989.1 3077 Missense Mutation CGG,TGG R859W XP_016867478.1

View Full Product Details