Product Details

SNP ID
rs201338701
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:150692322 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAACTTGCTCCTCACAGGACCACA[C/T]GCAAAGGGCCAATGTGCCAGCAGAT
Phenotype
MIM: 608085
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GIMAP2 PubMed Links

Gene Details

Gene
GIMAP2
Gene Name
GTPase, IMAP family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015660.2 124 Silent Mutation CAC,CAT H12H NP_056475.1

View Full Product Details