Product Details

SNP ID
rs201193467
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:43877369 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGACTTGCCCGTGCCTGGCACCCC[A/G]ACGGTTGACAGAACCACCAGCCTTG
Phenotype
MIM: 611986 MIM: 610337
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MRPS24 PubMed Links

Gene Details

Gene
MRPS24
Gene Name
mitochondrial ribosomal protein S24
There are no transcripts associated with this gene.

Gene
URGCP
Gene Name
upregulator of cell proliferation
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077663.2 2583 Silent Mutation GTC,GTT V698V NP_001071131.1
NM_001077664.2 2583 Silent Mutation GTC,GTT V655V NP_001071132.1
NM_001290075.1 2583 Silent Mutation GTC,GTT V655V NP_001277004.1
NM_001290076.1 2583 Silent Mutation GTC,GTT V655V NP_001277005.1
NM_017920.4 2583 Silent Mutation GTC,GTT V689V NP_060390.3
Gene
URGCP-MRPS24
Gene Name
URGCP-MRPS24 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204871.1 2583 Intron NP_001191800.1

View Full Product Details