Product Details

SNP ID
rs201269935
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:16860571 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTCCAGCTATGTCAGCTCTAACT[C/G]TTAAAGAAGGGTCTGTCAAGGAAAA
Phenotype
MIM: 609482
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
AGR3 PubMed Links

Gene Details

Gene
AGR3
Gene Name
anterior gradient 3, protein disulphide isomerase family member
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_176813.4 432 Missense Mutation ACA,AGA T127R NP_789783.1
XM_005249627.2 432 Missense Mutation ACA,AGA T127R XP_005249684.1
XM_011515152.2 432 Missense Mutation ACA,AGA T127R XP_011513454.1

View Full Product Details