Product Details
- SNP ID
-
rs201163996
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:100573892 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGAGATCTTCTTCCTTTATGTTCCC[A/C]AAAACGCAGTCATCTTTAGCTATCT
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
SNX31
PubMed Links
Gene Details
- Gene
- SNX31
- Gene Name
- sorting nexin 31
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_152628.3 |
1527 |
Missense Mutation |
TTG,TTT |
L432F |
NP_689841.3 |
XM_005250815.3 |
1527 |
Missense Mutation |
TTG,TTT |
L333F |
XP_005250872.1 |
XM_011516899.1 |
1527 |
Missense Mutation |
TTG,TTT |
L366F |
XP_011515201.1 |
XM_017013153.1 |
1527 |
Missense Mutation |
TTG,TTT |
L431F |
XP_016868642.1 |
XM_017013154.1 |
1527 |
Missense Mutation |
TTG,TTT |
L431F |
XP_016868643.1 |
XM_017013155.1 |
1527 |
Missense Mutation |
TTG,TTT |
L430F |
XP_016868644.1 |
XM_017013156.1 |
1527 |
Missense Mutation |
TTG,TTT |
L413F |
XP_016868645.1 |
XM_017013157.1 |
1527 |
Missense Mutation |
TTG,TTT |
L394F |
XP_016868646.1 |
XM_017013158.1 |
1527 |
Missense Mutation |
TTG,TTT |
L393F |
XP_016868647.1 |
XM_017013159.1 |
1527 |
Missense Mutation |
TTG,TTT |
L375F |
XP_016868648.1 |
XM_017013160.1 |
1527 |
Missense Mutation |
TTG,TTT |
L364F |
XP_016868649.1 |
XM_017013161.1 |
1527 |
Missense Mutation |
TTG,TTT |
L349F |
XP_016868650.1 |
XM_017013162.1 |
1527 |
Missense Mutation |
TTG,TTT |
L326F |
XP_016868651.1 |
XM_017013163.1 |
1527 |
Missense Mutation |
TTG,TTT |
L227F |
XP_016868652.1 |
XM_017013164.1 |
1527 |
Intron |
|
|
XP_016868653.1 |
XM_017013165.1 |
1527 |
Intron |
|
|
XP_016868654.1 |
View Full Product Details