Product Details

SNP ID
rs200566009
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:99891958 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATAGCACGAATGCTACAGCCATAT[G/T]ATTTCGCAGACGCCTGGCCAGAAGG
Phenotype
MIM: 124090
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
COX6C PubMed Links

Gene Details

Gene
COX6C
Gene Name
cytochrome c oxidase subunit 6C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004374.3 471 Missense Mutation NP_004365.1
XM_017013020.1 471 Intron XP_016868509.1

View Full Product Details