Product Details

SNP ID
rs200809010
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:122820373 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAACTTGACAGCTGGGTACTCTGC[A/G]GCAAGGCAGATCATGCAACCATTCA
Phenotype
MIM: 604421
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PDCL PubMed Links

Gene Details

Gene
PDCL
Gene Name
phosducin like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005388.4 585 Silent Mutation GCC,GCT A206A NP_005379.3
XM_017014785.1 585 Silent Mutation GCC,GCT A141A XP_016870274.1

View Full Product Details