Product Details

SNP ID
rs201280945
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:32541633 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACTAAGTGTGGCAATATCACATTC[C/T]TTGTCCAGCACACCAAATTCAGCTT
Phenotype
MIM: 609507
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TOPORS PubMed Links

Gene Details

Gene
TOPORS
Gene Name
TOP1 binding arginine/serine rich protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195622.1 2889 Silent Mutation AAA,AAG K899K NP_001182551.1
NM_005802.4 2889 Silent Mutation AAA,AAG K964K NP_005793.2
Gene
TOPORS-AS1
Gene Name
TOPORS antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details