Product Details
- SNP ID
-
rs199750679
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:78240151 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAGTGTGCTAAAGAACCTGGAGCTA[A/G]ATGGACTAATTCTGAGAGAGAGGGA
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CEP78
PubMed Links
Gene Details
- Gene
- CEP78
- Gene Name
- centrosomal protein 78
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001098802.1 |
671 |
Missense Mutation |
AAT,GAT |
N128D |
NP_001092272.1 |
NM_032171.1 |
671 |
Missense Mutation |
AAT,GAT |
N128D |
NP_115547.1 |
XM_005252263.4 |
671 |
Missense Mutation |
AAT,GAT |
N128D |
XP_005252320.1 |
XM_005252265.2 |
671 |
Missense Mutation |
AAT,GAT |
N128D |
XP_005252322.1 |
XM_005252266.4 |
671 |
Missense Mutation |
AAT,GAT |
N41D |
XP_005252323.1 |
XM_017015192.1 |
671 |
Missense Mutation |
AAT,GAT |
N128D |
XP_016870681.1 |
XM_017015193.1 |
671 |
Missense Mutation |
AAT,GAT |
N128D |
XP_016870682.1 |
XM_017015194.1 |
671 |
Missense Mutation |
AAT,GAT |
N128D |
XP_016870683.1 |
XM_017015195.1 |
671 |
Missense Mutation |
AAT,GAT |
N128D |
XP_016870684.1 |
XM_017015196.1 |
671 |
Missense Mutation |
AAT,GAT |
N41D |
XP_016870685.1 |
View Full Product Details