Product Details

SNP ID
rs200982109
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:130384469 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATCAAACTCTGAGCTGGAGATAG[G/T]GACGATGTGACAGGAACTTTCCCTG
Phenotype
MIM: 300513
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
GPR119 PubMed Links

Gene Details

Gene
GPR119
Gene Name
G protein-coupled receptor 119
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178471.2 1069 Missense Mutation ACT,CCT T327P NP_848566.1

View Full Product Details