Product Details

SNP ID
rs201035309
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:49117600 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGGGATATCTTGGTTCCTTACCTT[A/C]CCATAGAGGCCTCGGTGAGGGCCAG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
GPKOW PubMed Links

Gene Details

Gene
GPKOW
Gene Name
G-patch domain and KOW motifs
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015698.5 856 Silent Mutation GGG,GGT G259G NP_056513.2
XM_017029415.1 856 Silent Mutation GGG,GGT G274G XP_016884904.1

View Full Product Details